Learn More
Grainger Hose Adapter 3/4 NPTF 20 mm
Catalog No. 502765883
Encompass_Preferred
The Grainger 3/4" NPTF Male x 20mm Female Brass Hose Adapter is made of brass for durability. It has a 3/4" NPTF male thread on one end to connect to NPTF threaded pipe, and a 20mm female thread on the other end to connect to a 20mm male threaded hose fitting. This adapter allows you to connect a 20mm hose to 3/4" NPTF threaded pipe. It has a maximum pressure rating of up to 232 PSI and temperature range of -40°F to 250°F. The dimensions are 1-1/4" length x 1" hex. This adapter can be used for hydraulic applications to adapt hose sizes.
Catalog No. 50-276-5883 Supplier Grainger Supplier No. 16P966
The Grainger 3/4" NPTF Male x 20mm Female Brass Hose Adapter is made of brass for durability. It has a 3/4" NPTF male thread on one end to connect to NPTF threaded pipe, and a 20mm female thread on the other end to connect to a 20mm male threaded hose fitting. This adapter allows you to connect a 20mm hose to 3/4" NPTF threaded pipe. It has a maximum pressure rating of up to 232 PSI and temperature range of -40°F to 250°F. The dimensions are 1-1/4" length x 1" hex. This adapter can be used for hydraulic applications to adapt hose sizes.