Learn More
Grainger Hose Adapter 3/4 NPTF 22 mm
Catalog No. 502767048
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5,000 psi and temperature range of -40°F to 212°F. This adapter connects a 3/4" hydraulic hose to 3/4" JIC fittings. It is designed for use in hydraulic applications where flexibility is needed. The adapter allows the hose to swivel and move freely while maintaining a leak-free connection. It can be used with most hydraulic fluids including oils, water glycols, water/oil emulsions, and synthetic fluids. This adapter provides a versatile solution for routing hydraulic hoses in tight spaces.
Catalog No. 50-276-7048 Supplier Grainger Supplier No. 16P967
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5,000 psi and temperature range of -40°F to 212°F. This adapter connects a 3/4" hydraulic hose to 3/4" JIC fittings. It is designed for use in hydraulic applications where flexibility is needed. The adapter allows the hose to swivel and move freely while maintaining a leak-free connection. It can be used with most hydraulic fluids including oils, water glycols, water/oil emulsions, and synthetic fluids. This adapter provides a versatile solution for routing hydraulic hoses in tight spaces.