Learn More
Grainger Hose Adapter 3/4 NPTF 26 mm
Catalog No. 502765881
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/4" NPTF to 26 mm hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to +212°F. This adapter allows you to connect 3/4" NPTF threaded ports to 26 mm hydraulic hose. It is manufactured in the USA by Adaptall.
Catalog No. 50-276-5881 Supplier Grainger Supplier No. 16P969
The ADAPTALL Hydraulic Hose Adapter is a 3/4" NPTF to 26 mm hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to +212°F. This adapter allows you to connect 3/4" NPTF threaded ports to 26 mm hydraulic hose. It is manufactured in the USA by Adaptall.