Learn More
Grainger Hose Adapter 3/4 NPTF 33 mm
Catalog No. 502765250
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 inch hose to 1/2 inch pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250 degrees Fahrenheit. The adapter is made of forged brass for durability and has a hex head for easy tightening. It can be used for air, water, oil, and inert gas applications. The adapter allows the connection of 3/4 inch inside diameter hose to 1/2 inch nominal pipe thread fittings.
Catalog No. 50-276-5250 Supplier Grainger Supplier No. 16P971
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 inch hose to 1/2 inch pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250 degrees Fahrenheit. The adapter is made of forged brass for durability and has a hex head for easy tightening. It can be used for air, water, oil, and inert gas applications. The adapter allows the connection of 3/4 inch inside diameter hose to 1/2 inch nominal pipe thread fittings.