Learn More
Grainger Hose Adapter 3/8 NPTF 3/4 NPTF
Catalog No. 502764517
Encompass_Preferred
The Grainger 3/8" NPTF Female x 3/4" NPTF Male Hose Adapter is made from brass for durability. It allows you to connect two hoses with different thread sizes. This adapter has a maximum pressure rating of 3000 PSI and temperature rating of 250°F. It measures 1-1/2" in length and weighs 0.1 lbs. The Grainger hose adapter converts between 3/8" NPT female threads on one end and 3/4" NPT male threads on the opposite end.
Catalog No. 50-276-4517 Supplier Grainger Supplier No. 16P915
The Grainger 3/8" NPTF Female x 3/4" NPTF Male Hose Adapter is made from brass for durability. It allows you to connect two hoses with different thread sizes. This adapter has a maximum pressure rating of 3000 PSI and temperature rating of 250°F. It measures 1-1/2" in length and weighs 0.1 lbs. The Grainger hose adapter converts between 3/8" NPT female threads on one end and 3/4" NPT male threads on the opposite end.