Learn More
Grainger Hose Adapter 5/16 NPTF 1/4 NPTF
Catalog No. 502764521
Encompass_Preferred
The Grainger Approved Hose Adapter is a 1/4" NPT male x 1/4" barb fitting made of brass. It can be used to connect 1/4" NPT threaded pipe to a 1/4" inside diameter hose. The maximum pressure rating is 3000 psi and the maximum temperature rating is 250°F. The fitting has a hex shaped body for easy tightening with a wrench. It can be used with air, water, oil and inert gas applications. The fitting is made in the USA.
Catalog No. 50-276-4521 Supplier Grainger Supplier No. 16P924
The Grainger Approved Hose Adapter is a 1/4" NPT male x 1/4" barb fitting made of brass. It can be used to connect 1/4" NPT threaded pipe to a 1/4" inside diameter hose. The maximum pressure rating is 3000 psi and the maximum temperature rating is 250°F. The fitting has a hex shaped body for easy tightening with a wrench. It can be used with air, water, oil and inert gas applications. The fitting is made in the USA.