Learn More
Accuflow Systems Saint Gobain P1SB Hose Assembly, 1 in. ID, 1 ea
Supplier:  Accuflow Systems 16P1SBTC16TC16SEPL98
Encompass_Preferred
Saint Gobain P1SB hose assemblies are designed for the transfer of high purity products in pharmaceutical, food, and cosmetic industries. These platinum-cured braided silicone hoses offer excellent resistance to temperature extremes and chemical attack, ensuring reliable performance in critical process applications.
  • Biopharmaceutical grade platinum-cured silicone
  • Resists temperature extremes, compression set, chemical attack, moisture, and environmental exposure
  • May be washed with any mild detergents
  • Sterilizable by autoclave, CIP, SIP, and gamma irradiation
  • Hose traceable with serial and lot numbers
Catalog No. NC2873712