Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Insert Fitting, PVDF, 1/2 in
Catalog No. NC2683164
Encompass_Preferred
Saint-Gobain Pure-Fit SIB insert fitting in PVDF provides a smooth inner bore transition between flexible tubing and Tri-Clamp process connections, minimizing dead volume and reducing sites for particle entrapment. The fitting is intended for aseptic fluid transfer and sanitary bioprocess assemblies where cleanliness and low particulate hold-up are required.
  • PVDF construction for chemical compatibility and low extractables.
  • Smooth inner bore design reduces dead volume and particle entrapment.
  • Nominal size 1/2 in, compatible with 1/2 in tubing.
  • Connections: barb x Tri-Clamp for sanitary process integration.
  • Maximum pressure 250 psi for typical process applications.
  • Suitable for aseptic fluid transfer and bioprocess assemblies.
Catalog No. NC2683164 Supplier Accuflow Systems Supplier No. PFLS500PVDF
Saint-Gobain Pure-Fit SIB insert fitting in PVDF provides a smooth inner bore transition between flexible tubing and Tri-Clamp process connections, minimizing dead volume and reducing sites for particle entrapment. The fitting is intended for aseptic fluid transfer and sanitary bioprocess assemblies where cleanliness and low particulate hold-up are required.
  • PVDF construction for chemical compatibility and low extractables.
  • Smooth inner bore design reduces dead volume and particle entrapment.
  • Nominal size 1/2 in, compatible with 1/2 in tubing.
  • Connections: barb x Tri-Clamp for sanitary process integration.
  • Maximum pressure 250 psi for typical process applications.
  • Suitable for aseptic fluid transfer and bioprocess assemblies.