Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD160112 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/16"" ID, 1/8"" Flow, Acetal
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0636409
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. NC1634365