Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD160412 Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/4"" ID, 1/8"" Flow, PP
Catalog No. 50628217
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-217 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636415
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.