Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD160212 Quick-Disconnect Panel-Mount Hose Barb Fitting Body; 1/8"" Tube ID, PP
Catalog No. NC1633271
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. NC1633271 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636410
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.