Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLC17004 Quick-Disconnect Hose Barb Body; Straight-through, 1/4"" ID, 1/4"" Flow, Acetal
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0636151
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-008