Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD170412 Quick-Disconnect Hose Barb Fitting Body; Valved, 1/4"" Tube ID, 1/8"" Flow Size
Catalog No. 50628231
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-231 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636425
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.