Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD170212 Quick-Disconnect Hose Barb Fitting Body; Valved, 1/8"" Tube ID, 1/8"" Flow Size
Catalog No. 50628225
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-225 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636420
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.