Learn More
Cole-Parmer Edwards C105-14-645 Vacuum Pump Fittings, aluminum adapter, NW 25 by 1/2 to 3/8" hose barb
Catalog No. NC0243823 Shop All Cole-Parmer Products
Encompass_Preferred
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .
Catalog No. NC0243823 Supplier Cole-Parmer Supplier No. 3140169
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .