Learn More
Cole-Parmer Hose Barb Fitting, Tee Union, Medical Nylon, Cleanroom Packed, 3/32" ID; 10/Pk
Catalog No. 502558146 Shop All Cole-Parmer Products
Encompass_Preferred
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These medical nylon fittings are made from the same resin as natural nylon but with FDA-compliant white coloring. They offer good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Catalog No. 50-255-8146 Supplier Cole-Parmer Supplier No. 5011081
Use these high-quality PVC-free fittings in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These medical nylon fittings are made from the same resin as natural nylon but with FDA-compliant white coloring. They offer good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.