Learn More
Cole-Parmer Hose Barb Fitting, Tee Union, Stainless Steel, 1/8" ID; 10/Pk
Supplier:  Cole-Parmer 5011100
Encompass_Preferred
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. Use the tee unions to combine or split fluid flow. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
Catalog No. 50-255-8128