Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) HFCD22612 Quick-disconnect fittings, Valved hose barbs inserts, PP, 3/8""
Catalog No. 50627728
Encompass
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.
Catalog No. 50-627-728 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3130345
High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately). Plug insert into coupling body to connect¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 SS latch springs and EPDM seals.