Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD2200412 Quick-Disconnect Hose Barb Insert; Valved, 1/4"" Tube ID, 1/4"" Flow Size, PP
Catalog No. 50628103
Encompass
CPC (Colder) PLCD2200412 Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/4" Flow Size, PP
Catalog No. 50-628-103 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636480
CPC (Colder) PLCD2200412 Quick-Disconnect Hose Barb Insert; Valved, 1/4" Tube ID, 1/4" Flow Size, PP