Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PMCD220212 Quick-Disconnect Hose Barb Insert; Valved, 1/8"" Tube ID, 1/8"" Flow Size, PP
Catalog No. 50628249
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-249 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636440
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.