Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) PLCD22006 Quick-Disconnect Hose Barb Insert; Valved, 3/8"" Tube ID, 1/4"" Flow Size, Acetal
Catalog No. 50628107
Encompass
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.
Catalog No. 50-628-107 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636085
Standard flow quick-disconnect couplings require a body and an insert of the same flow size. Plug insert into body to connect fitting¿fitting halves lock into place immediately. To disconnect fitting halves, simply press the thumb latch on the coupling body.