Learn More
Grainger ProLine Series Hose Bibb, Brass, 1/2 in. FIP x 3/4 in. MHT, 1/4 Turn, 1 ea
Catalog No. 502793380
Encompass_Preferred
This brass hose bibb provides a reliable outdoor water supply point for residential and commercial settings. The valve features an angled body design that prevents hose kinking and flow restriction, ensuring efficient performance.
  • Forged one-piece brass body for leak resistance
  • Easy quarter-turn handle for convenient on/off operation
  • Teflon seats and chrome-plated brass ball
  • Maximum working pressure of 125 PSI
  • Operating temperature range of 40°F to 160°F
  • 1/2 in. FIP inlet and 3/4 in. MHT outlet
  • Compliant with low-lead standards
  • Conforms to ASME A112.18.1/CSA B125.1 standards
Catalog No. 50-279-3380 Supplier Grainger Supplier No. 1APY2
This brass hose bibb provides a reliable outdoor water supply point for residential and commercial settings. The valve features an angled body design that prevents hose kinking and flow restriction, ensuring efficient performance.
  • Forged one-piece brass body for leak resistance
  • Easy quarter-turn handle for convenient on/off operation
  • Teflon seats and chrome-plated brass ball
  • Maximum working pressure of 125 PSI
  • Operating temperature range of 40°F to 160°F
  • 1/2 in. FIP inlet and 3/4 in. MHT outlet
  • Compliant with low-lead standards
  • Conforms to ASME A112.18.1/CSA B125.1 standards