Learn More
Grainger Approved Vendor Hose Bibb, Quarter-Turn, Brass, 3/4 in, 1 ea
Catalog No. 502793445
Encompass_Preferred
This hose bibb provides a reliable water source for exterior building applications, including garden irrigation and vehicle maintenance. Designed for ease of use, this unit features a quarter-turn mechanism for quick activation.
  • Features a durable brass body with an aluminum handle
  • Quarter-turn operation for efficient water control
  • Designed with a no-kink configuration
  • Compatible with 3/4 in. MNPT inlet and 3/4 in. GHT outlet connections
  • Rated for a maximum pressure of 125 psi and a maximum temperature of 180 °F
  • Complies with ANSI B1.20.1, ANSI B1.20.7, and ASME A112.18.1/CSA B125.1 standards
  • Certified to ANSI/NSF 61-9
Catalog No. 50-279-3445 Supplier Grainger Supplier No. 1APY1
This hose bibb provides a reliable water source for exterior building applications, including garden irrigation and vehicle maintenance. Designed for ease of use, this unit features a quarter-turn mechanism for quick activation.
  • Features a durable brass body with an aluminum handle
  • Quarter-turn operation for efficient water control
  • Designed with a no-kink configuration
  • Compatible with 3/4 in. MNPT inlet and 3/4 in. GHT outlet connections
  • Rated for a maximum pressure of 125 psi and a maximum temperature of 180 °F
  • Complies with ANSI B1.20.1, ANSI B1.20.7, and ASME A112.18.1/CSA B125.1 standards
  • Certified to ANSI/NSF 61-9