Learn More
Grainger Approved Vendor Hose Bibb, Brass, Quarter-Turn, 3/4 in, 1 ea
Catalog No. 502793362
Encompass_Preferred
This hose bibb is designed for reliable water flow control and performance in various applications. It features a robust construction suitable for demanding service requirements and is engineered for ease of use with a quarter-turn handle mechanism.
  • Brass body and trim construction for enhanced durability
  • Aluminum handle material for lightweight operation
  • Quarter-turn design for quick and efficient flow control
  • Pressure rating of 125 psi
  • Maximum temperature rating of 180 °F
  • Compliant with ANSI B1.20.1, ANSI B1.20.7, and ASME A112.18.1/CSA B125.1 standards
  • Low lead compliant and IAPMO NSI/NSF 61 approved
  • 3/4 in. MNPT inlet and 3/4 in. GHT outlet connection sizes
Catalog No. 50-279-3362 Supplier Grainger Supplier No. 1APV8
This hose bibb is designed for reliable water flow control and performance in various applications. It features a robust construction suitable for demanding service requirements and is engineered for ease of use with a quarter-turn handle mechanism.
  • Brass body and trim construction for enhanced durability
  • Aluminum handle material for lightweight operation
  • Quarter-turn design for quick and efficient flow control
  • Pressure rating of 125 psi
  • Maximum temperature rating of 180 °F
  • Compliant with ANSI B1.20.1, ANSI B1.20.7, and ASME A112.18.1/CSA B125.1 standards
  • Low lead compliant and IAPMO NSI/NSF 61 approved
  • 3/4 in. MNPT inlet and 3/4 in. GHT outlet connection sizes