Learn More
Grainger B&K LLC Hose Bibb, Brass, 1/2 in MIP, 3/4 in MHT, 1 ea
Catalog No. 502793135
Encompass_Preferred
This hose bibb provides reliable flow control for outdoor residential or commercial water applications. Constructed with a durable brass body and featuring a chrome-plated brass ball for enhanced corrosion resistance, this valve is designed for long-lasting performance.
  • Quarter-turn handle for easy and efficient operation
  • Durable brass construction for extended service life
  • Chrome-plated brass ball for superior corrosion resistance
  • Rust-resistant handle design
  • Complies with ASME A112.18.1/CSA B125.1 standards
  • 125 PSI maximum working pressure
  • Rated for operating temperatures up to 180°F
  • 1/2 in. MIP inlet and 3/4 in. MHT outlet
Catalog No. 50-279-3135 Supplier Grainger Supplier No. 1APV7
This hose bibb provides reliable flow control for outdoor residential or commercial water applications. Constructed with a durable brass body and featuring a chrome-plated brass ball for enhanced corrosion resistance, this valve is designed for long-lasting performance.
  • Quarter-turn handle for easy and efficient operation
  • Durable brass construction for extended service life
  • Chrome-plated brass ball for superior corrosion resistance
  • Rust-resistant handle design
  • Complies with ASME A112.18.1/CSA B125.1 standards
  • 125 PSI maximum working pressure
  • Rated for operating temperatures up to 180°F
  • 1/2 in. MIP inlet and 3/4 in. MHT outlet