Learn More
AVANTOR MASTERFLEX (VWR LLC) Acetal Copolymer Hose Clamps, White, 0.859"" x 0.989"", 100/pack
Catalog No. NC1397347
Encompass
Acetal Copolymer Hose Clamps, White, 0.859" x 0.989", 100/pack, Recommended for continuous use from ambient temperature to 200¿F (93¿C). Clamps are nonconducting and stable in a wide variety of chemicals. Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.
Catalog No. NC1397347 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0683220
Acetal Copolymer Hose Clamps, White, 0.859" x 0.989", 100/pack, Recommended for continuous use from ambient temperature to 200¿F (93¿C). Clamps are nonconducting and stable in a wide variety of chemicals. Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.