Learn More
Grainger Hose Clamp Kit 4 Band L 1/2 W PK10
Catalog No. 502767427
Encompass_Preferred
The Grainger 16P340 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It has a cast iron body with standard mounting dimensions and comes with a 1 year warranty. The motor can provide 0.56 kW of mechanical power from electrical input and is suitable for various industrial and commercial applications requiring up to 3/4 horsepower.
Catalog No. 50-276-7427 Supplier Grainger Supplier No. 16P340
The Grainger 16P340 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It has a cast iron body with standard mounting dimensions and comes with a 1 year warranty. The motor can provide 0.56 kW of mechanical power from electrical input and is suitable for various industrial and commercial applications requiring up to 3/4 horsepower.