Learn More
Grainger Manitowoc Hose Clamp, Snap, 0.799 in. to 0.899 in. ID, 2/Pk.
Catalog No. 502795492
Encompass_Preferred
This snap hose clamp is designed for secure fastening in refrigeration compartment applications, including use within plastic drain lines for ice machines. It features an inner diameter of 0.799 in. to 0.899 in. and is manufactured from durable plastic material.
  • Snap-style design for efficient installation.
  • Compatible with a wide range of ice machine models and cabinet systems.
  • Designed for use in foodservice and beverage industry equipment.
  • Supplied in a pack of 2 units.
Catalog No. 50-279-5492 Supplier Grainger Supplier No. 19XL47
This snap hose clamp is designed for secure fastening in refrigeration compartment applications, including use within plastic drain lines for ice machines. It features an inner diameter of 0.799 in. to 0.899 in. and is manufactured from durable plastic material.
  • Snap-style design for efficient installation.
  • Compatible with a wide range of ice machine models and cabinet systems.
  • Designed for use in foodservice and beverage industry equipment.
  • Supplied in a pack of 2 units.