Learn More
Ibidi USA Hose Clip
SDP
Catalog No. NC1849424
Encompass
A hose clip used for temporary or permanent closing of silicone tubes. Allows easy closing and re-opening of silicone tubing. Ideal for all flow applications
This item is not returnable. View return policy
Catalog No. NC1849424 Supplier Ibidi USA Supplier No. 10821
A hose clip used for temporary or permanent closing of silicone tubes. Allows easy closing and re-opening of silicone tubing. Ideal for all flow applications