Learn More
Chemglass Life Sciences Thread size: GL-18, Style: Bent, Hose OD (mm): 10.5, Hose ID (mm): 7, Length (mm): 65
SDP
Catalog No. CG198H16
Encompass_Preferred
Removable hose connections, high temperature, Thread size: GL-18, Style: Bent, Hose OD (mm): 10.5, Hose ID (mm): 7, Length (mm): 65. PFA hose connections with a Viton rubber seal for use with GL-14 or GL-18 glass screw threads. Enables the connection of cooling or vacuum hose by simply screwing either style (straight or bent) onto the glass thread instead of forcing tubing over a serrated glass connector. Connection will withstand temperatures up to 250¿C.*** CAPS SOLD SEPARATELY. SEE CG-195-H ***
Catalog No. CG198H16 Supplier Chemglass Life Sciences Supplier No. CG198H16
Removable hose connections, high temperature, Thread size: GL-18, Style: Bent, Hose OD (mm): 10.5, Hose ID (mm): 7, Length (mm): 65. PFA hose connections with a Viton rubber seal for use with GL-14 or GL-18 glass screw threads. Enables the connection of cooling or vacuum hose by simply screwing either style (straight or bent) onto the glass thread instead of forcing tubing over a serrated glass connector. Connection will withstand temperatures up to 250¿C.*** CAPS SOLD SEPARATELY. SEE CG-195-H ***