Learn More
Ace Glass, Inc. Hose connector, angled, GL14, 8mm OD, polypropylene with seal
SDP
Catalog No. NC9085133
Encompass
Duran, polypropylene hose connector with silicone seal. All are 8mm OD, 4mm ID. Use in conjunction with our 7621 open top cap Duran product family.
Catalog No. NC9085133 Supplier Ace Glass, Inc. Supplier No. 762320
Duran, polypropylene hose connector with silicone seal. All are 8mm OD, 4mm ID. Use in conjunction with our 7621 open top cap Duran product family.