Learn More
Grainger Westward Grease Gun Hose, Polyurethane, 12 in., 1/8 in. NPT, 1 ea.
Catalog No. 502754106
Encompass_Preferred
This flexible polyurethane hose extension facilitates lubrication in confined spaces by allowing grease guns to navigate around obstacles. Designed for reliability in demanding conditions, it features an integrated anti-kink spring guard and durable steel fittings.
  • Flexible polyurethane construction
  • 13,775 psi burst pressure
  • 5,800 psi maximum operating pressure
  • 12 in. overall length
  • 1/8 in. NPT end threads
  • Includes spring guard and steel fittings
  • Corrosion, impact, and UV resistant
  • Compatible with manual grease guns
Catalog No. 50-275-4106 Supplier Grainger Supplier No. 15F204
This flexible polyurethane hose extension facilitates lubrication in confined spaces by allowing grease guns to navigate around obstacles. Designed for reliability in demanding conditions, it features an integrated anti-kink spring guard and durable steel fittings.
  • Flexible polyurethane construction
  • 13,775 psi burst pressure
  • 5,800 psi maximum operating pressure
  • 12 in. overall length
  • 1/8 in. NPT end threads
  • Includes spring guard and steel fittings
  • Corrosion, impact, and UV resistant
  • Compatible with manual grease guns