Learn More
Grainger Hose Fitting Sleeve 1/2 In
Catalog No. 502749750
Encompass_Preferred
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting used for connecting tubing in fluid power systems. It has a 0.25 inch tube size and is made from brass for durability and corrosion resistance. This compression sleeve has a maximum pressure rating of 5000 psi and temperature range of -65 to 300 degrees Fahrenheit. The fitting accepts 0.25 inch OD tubing and uses a ferrule and nut to form a secure, leak-free connection when properly installed. This brass compression sleeve fitting is ideal for use in hydraulic, pneumatic, lubrication, fuel and oil applications.
Catalog No. 50-274-9750 Supplier Grainger Supplier No. 14L793
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting used for connecting tubing in fluid power systems. It has a 0.25 inch tube size and is made from brass for durability and corrosion resistance. This compression sleeve has a maximum pressure rating of 5000 psi and temperature range of -65 to 300 degrees Fahrenheit. The fitting accepts 0.25 inch OD tubing and uses a ferrule and nut to form a secure, leak-free connection when properly installed. This brass compression sleeve fitting is ideal for use in hydraulic, pneumatic, lubrication, fuel and oil applications.