Learn More
Grainger Hose Fitting Sleeve 5/16 In
Catalog No. 502753123
Encompass_Preferred
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting for use with AQP hose assemblies. It has a working pressure of 3000 psi and temperature range of -65 to +275°F. This compression sleeve is made of brass per SAE J514, has a thread size of 9/16-18 UNF, and is designed for use with 9/16" ID hose. The sleeve measures 1.28" in length. This brass compression sleeve fitting allows easy assembly and provides a heavy duty seal. It is an ideal choice for hydraulic applications using Aeroquip hose.
Catalog No. 50-275-3123 Supplier Grainger Supplier No. 14L797
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting for use with AQP hose assemblies. It has a working pressure of 3000 psi and temperature range of -65 to +275°F. This compression sleeve is made of brass per SAE J514, has a thread size of 9/16-18 UNF, and is designed for use with 9/16" ID hose. The sleeve measures 1.28" in length. This brass compression sleeve fitting allows easy assembly and provides a heavy duty seal. It is an ideal choice for hydraulic applications using Aeroquip hose.