Learn More
Grainger Hose Fitting Sleeve 5/8 In
Catalog No. 502749801
Encompass_Preferred
The Aeroquip Compression Sleeve Brass is a fitting for joining tubing in fluid power systems. It features a one-piece design with an integral ferrule and nut, made from brass for corrosion resistance. With a pressure rating of 3000 psi and a temperature range of -65 to 300°F, it accommodates tube sizes from 1/8 to 1 inch. Known for high quality, Aeroquip by Eaton is used in industrial, mobile, and aerospace applications. This sleeve simplifies installations by reducing parts, offering an economical choice for secure tubing connections.
Catalog No. 50-274-9801 Supplier Grainger Supplier No. 14L794
The Aeroquip Compression Sleeve Brass is a fitting for joining tubing in fluid power systems. It features a one-piece design with an integral ferrule and nut, made from brass for corrosion resistance. With a pressure rating of 3000 psi and a temperature range of -65 to 300°F, it accommodates tube sizes from 1/8 to 1 inch. Known for high quality, Aeroquip by Eaton is used in industrial, mobile, and aerospace applications. This sleeve simplifies installations by reducing parts, offering an economical choice for secure tubing connections.