Learn More
Grainger Hose Fitting Sleeve 7/8 In
Catalog No. 502749751
Encompass_Preferred
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting used for joining tubing in fluid power systems. It has a one piece design with an integral ferrule and nut. The body is made of brass for corrosion resistance. It has a pressure rating of 3000 psi and temperature range of -65 to 300°F. The sleeve accommodates tubing sizes from 1/8" to 1/2" OD. This compression sleeve provides a strong, reusable seal for hydraulic and pneumatic tubing connections. It is an ideal choice for use in industrial, mobile, and marine applications.
Catalog No. 50-274-9751 Supplier Grainger Supplier No. 14L795
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting used for joining tubing in fluid power systems. It has a one piece design with an integral ferrule and nut. The body is made of brass for corrosion resistance. It has a pressure rating of 3000 psi and temperature range of -65 to 300°F. The sleeve accommodates tubing sizes from 1/8" to 1/2" OD. This compression sleeve provides a strong, reusable seal for hydraulic and pneumatic tubing connections. It is an ideal choice for use in industrial, mobile, and marine applications.