Learn More
Grainger Hose Fitting Sleeve 7/8 In
Catalog No. 502751455
Encompass_Preferred
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting used for creating leak-proof connections in fluid transfer applications. It has a 1/4" NPT female pipe thread on one end and a compression connection on the other end to connect to 1/4" OD tubing. The compression sleeve is made from brass for corrosion resistance. This fitting allows a tubing connection without flaring and can handle pressures up to 5000 psi. It is commonly used in hydraulic, pneumatic, lubrication, and industrial process systems.
Catalog No. 50-275-1455 Supplier Grainger Supplier No. 14L796
The Aeroquip Compression Sleeve Brass is a brass compression sleeve fitting used for creating leak-proof connections in fluid transfer applications. It has a 1/4" NPT female pipe thread on one end and a compression connection on the other end to connect to 1/4" OD tubing. The compression sleeve is made from brass for corrosion resistance. This fitting allows a tubing connection without flaring and can handle pressures up to 5000 psi. It is commonly used in hydraulic, pneumatic, lubrication, and industrial process systems.