Learn More
Perkin Elmer US LLC Flame Capillary Tubing Assembly for PerkinElmer AS-90/AS-90A/AS-90 Plus/AS-91/AS-93 Plus/AS-S10
Catalog No. 509042746
Encompass_Preferred
NexION 1000/2000 internal standard mixing tee is perfect for online addition of internal standard or online dilution. Achieves excellent mixing with minimal dead volume.
Catalog No. 50-904-2746 Supplier Perkin Elmer US LLC Supplier No. B3000157
NexION 1000/2000 internal standard mixing tee is perfect for online addition of internal standard or online dilution. Achieves excellent mixing with minimal dead volume.