Learn More
NewAge Industries Hose Holder
Description
Hose Holders by AdvantaPure™ is the ultimate solution for safe and efficient tubing and hose management in biopharmaceutical manufacturing facilities. Designed specifically to maintain the integrity of biopharmaceutical operations, the Hose Holder reduces tripping hazards, contamination, and tubing and hose damage during the transfer of liquids from one vessel to another. Made of high-quality materials, it is designed to secure tubing and hoses of various sizes and sits easily on any flat surface. Hose Holders by AdvantaPure helps improve cleanliness and organization by keeping tubing and hose off the ground and limiting tangles and kinking. Easy to assemble by aligning the raised notches on either side, it clicks in place to be used immediately with no tools needed for assembly or installation. The Hose Holder is an ideal solution for aiding safe and efficient fluid transfer by keeping your tubing and hose off the floor.
- Dimensions: 12 x 5 x 5 in. after assembly
- Available in cases of 20 disassembled Hose Holders that are double bagged
Specifications
Specifications
| Packaging Quantity | 20 Yellow Hose Holders |
| Product Type | Tube Holder |