Learn More
NewAge Industries Hose Holder
Catalog No. 014815452
Change view
Click to view available options
Packaging Quantity:
20 Orange Hose Holders
20 Yellow Hose Holders
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
01-481-5452 20 Orange Hose Holders
01-481-5430 20 Yellow Hose Holders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-481-5452 Supplier NewAge Industries Supplier No. HH1101000
Add to Cart
Add to Cart
Tubing and hose management solution for manufacturing. Reduces potential contamination from the floor as hose is being handled. Visible colors help reduce tripping hazards. Standard Hose Holder raw materials are REACH and EMEA/410/01 compliant. Easy to use, easy to clean and maintain.
Hose Holders by AdvantaPure™ is the ultimate solution for safe and efficient tubing and hose management in biopharmaceutical manufacturing facilities. Designed specifically to maintain the integrity of biopharmaceutical operations, the Hose Holder reduces tripping hazards, contamination, and tubing and hose damage during the transfer of liquids from one vessel to another. Made of high-quality materials, it is designed to secure tubing and hoses of various sizes and sits easily on any flat surface. Hose Holders by AdvantaPure helps improve cleanliness and organization by keeping tubing and hose off the ground and limiting tangles and kinking. Easy to assemble by aligning the raised notches on either side, it clicks in place to be used immediately with no tools needed for assembly or installation. The Hose Holder is an ideal solution for aiding safe and efficient fluid transfer by keeping your tubing and hose off the floor.
  • Dimensions: 12 x 5 x 5 in. after assembly
  • Available in cases of 20 disassembled Hose Holders that are double bagged
Specifications
Packaging Quantity 20 Orange Hose Holders
Product Type Tube Holder