Learn More
Grainger Hose Kit American Standard
Catalog No. 502766924
Encompass_Preferred
The American Standard Hose Kit Fits American Standard Toilets is a replacement fill valve hose kit for American Standard toilets. It includes a 7" braided stainless steel supply line with a 3/8" compression nut on angle stop end and a 5/8" nut with washer on the fill valve end. This hose kit is designed for use with most American Standard toilets and is made from durable stainless steel for longevity. The kit provides an easy way to replace a worn or leaking existing fill valve hose.
Catalog No. 50-276-6924 Supplier Grainger Supplier No. 16U031
The American Standard Hose Kit Fits American Standard Toilets is a replacement fill valve hose kit for American Standard toilets. It includes a 7" braided stainless steel supply line with a 3/8" compression nut on angle stop end and a 5/8" nut with washer on the fill valve end. This hose kit is designed for use with most American Standard toilets and is made from durable stainless steel for longevity. The kit provides an easy way to replace a worn or leaking existing fill valve hose.