Learn More
Grainger DEWALT Auto Retracting Air Hose Reel, 1/2 in. x 50 ft., All-Steel, 250 PSI, 1 ea.
Catalog No. 502786699
Encompass_Preferred
This double arm auto-retracting air hose reel features a robust all-steel construction designed for reliable industrial, contractor, or household use. The system includes a 1/2 in. x 50 ft. nitrile rubber hose, engineered to remain flexible in temperatures as low as -40°F and resistant to heat up to 150°F. With a maximum working pressure of 250 psi, this unit provides durable performance for demanding pneumatic applications.
  • Spring-powered drive for quick and automatic hose retraction
  • 12 position ratchet gearing lock for customizable hose length
  • Self-leveling system for easy winding
  • Professional grade dual-arm all-steel construction
  • Four-roller hose guide design to reduce wear
  • Kink and oil resistant hose
  • Includes 1/2 in. x 4 ft. lead-in air hose
Catalog No. 50-278-6699 Supplier Grainger Supplier No. 190R86
This double arm auto-retracting air hose reel features a robust all-steel construction designed for reliable industrial, contractor, or household use. The system includes a 1/2 in. x 50 ft. nitrile rubber hose, engineered to remain flexible in temperatures as low as -40°F and resistant to heat up to 150°F. With a maximum working pressure of 250 psi, this unit provides durable performance for demanding pneumatic applications.
  • Spring-powered drive for quick and automatic hose retraction
  • 12 position ratchet gearing lock for customizable hose length
  • Self-leveling system for easy winding
  • Professional grade dual-arm all-steel construction
  • Four-roller hose guide design to reduce wear
  • Kink and oil resistant hose
  • Includes 1/2 in. x 4 ft. lead-in air hose