Learn More
Grainger Coxreels Spring Return Hose Reel, Stainless Steel, 25 ft, 3/4 in I.D., 1 ea
Catalog No. 502766103
Encompass_Preferred
This spring-driven, heavy-duty hose reel is engineered from electro-polished stainless steel for reliable, non-corrosive performance in environments where rust, corrosion, and contamination are concerns, such as food and chemical processing. It is designed for use with air, water, or mild chemicals.
  • Solid, one-piece heavy-gauge stainless steel base and support post for maximum stability
  • Multi-position lock ratchet mechanism secures hose at the desired length
  • 90° full-flow NPT swivel inlet with non-toxic, food-grade nitrile seals
  • Enclosed, factory-tuned, and removable cartridge-style spring motor
  • Permanently lubricated precision bearings for smooth rotation
  • CNC-spun, ribbed discs with rolled edges for enhanced strength and safety
  • Maximum operating pressure of 300 psi
  • Operating temperature range of -40°F to 150°F
Catalog No. 50-276-6103 Supplier Grainger Supplier No. 16X548
This spring-driven, heavy-duty hose reel is engineered from electro-polished stainless steel for reliable, non-corrosive performance in environments where rust, corrosion, and contamination are concerns, such as food and chemical processing. It is designed for use with air, water, or mild chemicals.
  • Solid, one-piece heavy-gauge stainless steel base and support post for maximum stability
  • Multi-position lock ratchet mechanism secures hose at the desired length
  • 90° full-flow NPT swivel inlet with non-toxic, food-grade nitrile seals
  • Enclosed, factory-tuned, and removable cartridge-style spring motor
  • Permanently lubricated precision bearings for smooth rotation
  • CNC-spun, ribbed discs with rolled edges for enhanced strength and safety
  • Maximum operating pressure of 300 psi
  • Operating temperature range of -40°F to 150°F