Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence Luer fittings, 316 SS fittings, female luer lock x 1/16"" hose barb 41507-29
Catalog No. NC0552213
Encompass
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.
Catalog No. NC0552213 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150729
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.