Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence Luer fittings, 316 SS fittings, male luer lock x 1/16"" hose barb 41507-26
Catalog No. NC0552212
Encompass
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.
Catalog No. NC0552212 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150726
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.