Learn More
Chemglass Life Sciences Hosecock
SDP
Catalog No. 50872173
Encompass_Preferred
Hosecock. For accurate regulation or interruption of fluid flow, these precision machined hosecocks are made to resist rust and corrosion. Adjustment screw with oversized head permits accurate regulation with easy one-hand operation. Used for tubing up to 19mm, the hosecocks feature built-in side lugs for foot mounting and a tubing retainer screw.
Catalog No. 50-872-173 Supplier Chemglass Life Sciences Supplier No. CG924001
Hosecock. For accurate regulation or interruption of fluid flow, these precision machined hosecocks are made to resist rust and corrosion. Adjustment screw with oversized head permits accurate regulation with easy one-hand operation. Used for tubing up to 19mm, the hosecocks feature built-in side lugs for foot mounting and a tubing retainer screw.