Learn More
Medchemexpress LLC HY-N2244 5mg Medchemexpress, Hosenkoside M CAS:161016-51-9 Purity:>98%
SDP
Catalog No. 501933897
Encompass_Preferred
Medchemexpress, HY-N2244 5mg Hosenkoside M CAS:161016-51-9 Hosenkoside M is a baccharane glycoside isolated from the seeds of impatiens balsamina. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3897 Supplier Medchemexpress LLC Supplier No. HYN22445MG
Medchemexpress, HY-N2244 5mg Hosenkoside M CAS:161016-51-9 Hosenkoside M is a baccharane glycoside isolated from the seeds of impatiens balsamina. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.