Learn More
GUARDIAN EQUIPMENT 8' PVC Hose
Catalog No. NC1318228
Encompass_Preferred
8 ft. gray PVC hose with 3/8" MNPT swivel inlet and 3/8" MNPT outlet with handle thread (for use with dual headed units)
Catalog No. NC1318228 Supplier GUARDIAN EQUIPMENT Supplier No. AP350100096
8 ft. gray PVC hose with 3/8" MNPT swivel inlet and 3/8" MNPT outlet with handle thread (for use with dual headed units)