Learn More
Enterprise Technology Solutions Hospeco Exam Gloves,ExtendedCuff,Nitrile,8 mil,2XL,100/BX,10BX/CT,BE
SDP
Catalog No. 5000828056
Encompass_Preferred
Nitrile Exam Gloves are powder-free and designed to provide extra protection and stronger resistance to wear and tear while on the job. 8 mil thick gloves are durable and allow you to work while keeping your hands safe and protected. Nonsterile, latex-free gloves feature an extended cuff for a full 12" glove length and maximum protection. Ambidextrous design is perfect for use in industries such as aerospace, automotive, healthcare, industrial/manufacturing, education, government, food-processing, foodservice, retail, hospitality and more.
Catalog No. 50-008-28056 Supplier Enterprise Technology Solutions Supplier No. HOSN108EPF2XL
Add to Cart
Add to Cart
Nitrile Exam Gloves are powder-free and designed to provide extra protection and stronger resistance to wear and tear while on the job. 8 mil thick gloves are durable and allow you to work while keeping your hands safe and protected. Nonsterile, latex-free gloves feature an extended cuff for a full 12" glove length and maximum protection. Ambidextrous design is perfect for use in industries such as aerospace, automotive, healthcare, industrial/manufacturing, education, government, food-processing, foodservice, retail, hospitality and more.