Learn More
Enterprise Technology Solutions Hospeco Exam Gloves,Nitrile,Powder-Free,5 mil,Small,100/BX,BK
SDP
Catalog No. 5000828079
Encompass_Preferred
ProWorks Black Nitrile Powder-Free Exam Gloves are made from 100 percent synthetic nitrile polymer and are completely latex-free, powder-free and odor-free. At 5 mil thick, they provide added durability while remaining softer and more elastic than most other nitrile gloves of similar thickness. These gloves deliver superior resistance to punctures, tears and chemicals compared to latex gloves. Textured surface ensures a strong, reliable grip in both wet and dry applications. Designed for versatility, they are ideal for general-purpose, industrial or mechanical use, including construction, law enforcement, agriculture, beauty salons and more.
Catalog No. 50-008-28079 Supplier Enterprise Technology Solutions Supplier No. HOSGLN145FS
ProWorks Black Nitrile Powder-Free Exam Gloves are made from 100 percent synthetic nitrile polymer and are completely latex-free, powder-free and odor-free. At 5 mil thick, they provide added durability while remaining softer and more elastic than most other nitrile gloves of similar thickness. These gloves deliver superior resistance to punctures, tears and chemicals compared to latex gloves. Textured surface ensures a strong, reliable grip in both wet and dry applications. Designed for versatility, they are ideal for general-purpose, industrial or mechanical use, including construction, law enforcement, agriculture, beauty salons and more.